Learn More
Cayman Chemical KB-0742 (hydrochloride), Purity: ≥95%, 10 mg
SDP
Catalog No. 502942162
Encompass_Preferred
A Cdk9 inhibitor (IC50 = 6 nM); selective for Cdk9 over Cdk1-8 (IC50s = 397->10,000 nM) and Cdk12-19 (IC50s = 372->10,000 nM); induces cell cycle arrest at the G1 phase and apoptosis in BT-20, BT-549, MDA-MB-231, MT-3, and Hs 578T TNBC cells from 0.64-7.73 µM; reduces tumor volume in the MYC-amplified CTG-0437 PDX mouse model of TNBC at 60 mg/kg
Catalog No. 50-294-2162 Supplier Cayman Chemical Supplier No. 3977210 MG
A Cdk9 inhibitor (IC50 = 6 nM); selective for Cdk9 over Cdk1-8 (IC50s = 397->10,000 nM) and Cdk12-19 (IC50s = 372->10,000 nM); induces cell cycle arrest at the G1 phase and apoptosis in BT-20, BT-549, MDA-MB-231, MT-3, and Hs 578T TNBC cells from 0.64-7.73 µM; reduces tumor volume in the MYC-amplified CTG-0437 PDX mouse model of TNBC at 60 mg/kg