Learn More
SURE TECH DIAGNOSTIC ASSOC INC KB FETAL HEMOGLOBIN KT-ST101-B
Catalog No. NC0224654
Encompass_Preferred
Includes 2 bottles each 101-10 Fixing Solution, 101-20 Citrate Buffer, 101-30 Staining Solution. Kleihauer Betke Staining Kit - For the identification of Fetal Eurythrocytes in Maternal Blood
Catalog No. NC0224654 Supplier SURE TECH DIAGNOSTIC ASSOC INC Supplier No. ST 101B
Includes 2 bottles each 101-10 Fixing Solution, 101-20 Citrate Buffer, 101-30 Staining Solution. Kleihauer Betke Staining Kit - For the identification of Fetal Eurythrocytes in Maternal Blood