Learn More
TARGETMOL CHEMICALS INC KB NB 142-70 10MG
SDP
Catalog No. 502581034
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. kb NB 142-70 is a selective protein kinase D (PKD) inhibitor (IC50 values are 28.3 58.7 and 53.2 nM for PKD1 2 and 3 respectively). purity: 99%
Catalog No. 50-258-1034 Supplier TARGETMOL CHEMICALS INC Supplier No. T206210MG
Add to Cart
Add to Cart
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. kb NB 142-70 is a selective protein kinase D (PKD) inhibitor (IC50 values are 28.3 58.7 and 53.2 nM for PKD1 2 and 3 respectively). purity: 99%