Learn More
Cayman Chemical kb NB 142-70 50mg
SDP
Catalog No. 502979686
Encompass_Preferred
A selective PKD inhibitor (IC50s = 28.3, 58.7, and 53.2 nM for PKD1, 2, and 3, respectively); inhibits prostate cancer cell migration and invasion, as well as reduces wound healing in vitro.
Catalog No. 50-297-9686 Supplier Cayman Chemical Supplier No. 1800250 MG
A selective PKD inhibitor (IC50s = 28.3, 58.7, and 53.2 nM for PKD1, 2, and 3, respectively); inhibits prostate cancer cell migration and invasion, as well as reduces wound healing in vitro.