Learn More
Medchemexpress LLC KB-R7943 mesylate | 182004-65-5 | C17H21N3O6S2 | 50 MG
SDP
Catalog No. 5000376140
Encompass_Preferred
KB-R7943 mesylate is an inhibitor of the reverse Na+/Ca2+ exchanger (NCXrev) with an IC50 of 5.7±2.1 μM. It induces cancer cell death via JNK pathway activation and autophagic flux blockade. The compound also blocks NMDAR-mediated ion currents and inhibits NMDA-induced increases in cytosolic Ca2+. It can depolarize mitochondria and inhibit 2,4-dinitrophenol-stimulated respiration of cultured neurons. This product is a solid, white to yellow compound with a molecular weight of 427.50.
  • Inhibits reverse Na+/Ca2+ exchanger (NCXrev).
  • Activates JNK pathway and blocks autophagic flux to induce cancer cell death.
  • Blocks NMDAR-mediated ion currents and inhibits NMDA-induced increases in cytosolic Ca2+.
  • Depolarizes mitochondria and inhibits 2,4-dinitrophenol-stimulated respiration.
  • Inhibits IhERG and native IKr.
  • Solid form, white to yellow color, molecular weight of 427.50.
Catalog No. 50-003-76140 Supplier Medchemexpress LLC Supplier No. HY1541550MG
KB-R7943 mesylate is an inhibitor of the reverse Na+/Ca2+ exchanger (NCXrev) with an IC50 of 5.7±2.1 μM. It induces cancer cell death via JNK pathway activation and autophagic flux blockade. The compound also blocks NMDAR-mediated ion currents and inhibits NMDA-induced increases in cytosolic Ca2+. It can depolarize mitochondria and inhibit 2,4-dinitrophenol-stimulated respiration of cultured neurons. This product is a solid, white to yellow compound with a molecular weight of 427.50.
  • Inhibits reverse Na+/Ca2+ exchanger (NCXrev).
  • Activates JNK pathway and blocks autophagic flux to induce cancer cell death.
  • Blocks NMDAR-mediated ion currents and inhibits NMDA-induced increases in cytosolic Ca2+.
  • Depolarizes mitochondria and inhibits 2,4-dinitrophenol-stimulated respiration.
  • Inhibits IhERG and native IKr.
  • Solid form, white to yellow color, molecular weight of 427.50.