Learn More
TARGETMOL CHEMICALS INC KB-R7943 MESYLATE 25MG
SDP
Catalog No. 502576591
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KB-R7943 mesylate is a widely used inhibitor of the reverse Na+/Ca2+ exchanger (NCXrev) with IC50 of 5.7±2.1 uM. purity: 99%
Catalog No. 50-257-6591 Supplier TARGETMOL CHEMICALS INC Supplier No. T614425MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KB-R7943 mesylate is a widely used inhibitor of the reverse Na+/Ca2+ exchanger (NCXrev) with IC50 of 5.7±2.1 uM. purity: 99%