Learn More
Selleck Chemical LLC KB-R7943 mesylate S4643-5mg
SDP
Catalog No. 503110844
Encompass_Preferred
KB-R7943 mesylate is a widely used inhibitor of the reverse Na /Ca2 exchanger (NCX(rev)) with IC50 of 5 7 M KB-R7943 mesylate promotes prostate cancer cell death by activating the JNK pathway and blocking autophagic flux
Catalog No. 50-311-0844 Supplier Selleck Chemical LLC Supplier No. S46435MG
KB-R7943 mesylate is a widely used inhibitor of the reverse Na /Ca2 exchanger (NCX(rev)) with IC50 of 5 7 M KB-R7943 mesylate promotes prostate cancer cell death by activating the JNK pathway and blocking autophagic flux