Learn More
Cayman Chemical KB-R7943mesylate 5mg
SDP
Catalog No. 502970267
Encompass_Preferred
A selective inhibitor of the reverse mode of NCX1 (IC50 = 1.2-2.4 µM) that prevents intracellular sodium-dependent calcium uptake in whole cells; exhibits neuroprotection from glutamate-induced excitotoxicity; inhibits TRPC channels involved in calcium-dependent signal transduction
Catalog No. 50-297-0267 Supplier Cayman Chemical Supplier No. 163525 MG
A selective inhibitor of the reverse mode of NCX1 (IC50 = 1.2-2.4 µM) that prevents intracellular sodium-dependent calcium uptake in whole cells; exhibits neuroprotection from glutamate-induced excitotoxicity; inhibits TRPC channels involved in calcium-dependent signal transduction