Learn More
Medchemexpress LLC HY-129610 5mg , KB02-SLF CAS:2384184-40-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1296105MG
Encompass_Preferred
HY-129610 5mg KB02-SLF CAS: 2384184-40-9 KB02-SLF is a PROTAC-based nuclear FKBP12 degrader (molecular glue). KB02-SLF promotes nuclear FKBP12 degradation by covalently modifying DCAF16 (E3 ligase) and can improve the durability of protein degradation in biological systems. SLF binds ubiquitin E3 ligase ligand KB02 via a linker to form KB02-SLF. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0688