Learn More
Biolegend KIRAVIA Blue 520™ anti-mouse CD11c
Supplier:  Biolegend 117364
Encompass_Preferred
KIRAVIA Blue 520™ anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-0298