Learn More
AdipoGen alpha-Galactosylceramide Analog I (water soluble) [KBC-007] (100 µg)
SDP
Supplier:  AdipoGen AGCR13608C100
Encompass_Preferred
Chemical. CAS: 1037297-61-2. Formula: C46H92N2O9. MW: 817.2. Synthetic, soluble analog of alpha-galactosylceramide (Prod. No. AG-CN2-0013). Immunostimulant. Potent stimulator of natural killer T (NKT) cells. Specific ligand of the lipid-binding MHC class I-like protein CD1d in human and mouse NKT cells.
Catalog No. 50-205-3977