Learn More
Lonza Walkersville KBM Gold Keratinocyte Growth Basal Medium
Catalog No. NC0135933
Encompass
KBM Gold is a keratinocyte basal medium sold separately. This can be used with 1 kit of 192152 (keratinocyte cell growth medium SingleQuots Supplements and Growth Factors). The full KGM Gold BulletKit consisting of basal medium and supplements can be purchased together using the catalog number 192060.KGM Gold BulleKit is a new robust and versatile serum-free medium formulation for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures.Content:1 x Bottle Basal Medium 500 mL
Catalog No. NC0135933 Supplier Lonza Walkersville Supplier No. 00192151
KBM Gold is a keratinocyte basal medium sold separately. This can be used with 1 kit of 192152 (keratinocyte cell growth medium SingleQuots Supplements and Growth Factors). The full KGM Gold BulletKit consisting of basal medium and supplements can be purchased together using the catalog number 192060.KGM Gold BulleKit is a new robust and versatile serum-free medium formulation for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures.Content:1 x Bottle Basal Medium 500 mL