Learn More
Lonza Walkersville KBM Gold Medium w/o Ca Ph Red
Catalog No. 5000496233
Encompass
KBM Gold is a calcium-free keratinocyte basal medium sold separately This can be used with 1 kit of 192152 (Keratinocyte Cell Growth Medium SingleQuots Supplements and Growth Factors) The full KGM Gold Calcium Free BulletKit consisting of basal medium and supplements can be purchased together using the catalog number 00195769 KGM Gold BulleKit is a robust and versatile serum-free medium formulation for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures
Catalog No. 50-004-96233 Supplier Lonza Walkersville Supplier No. 00195130
KBM Gold is a calcium-free keratinocyte basal medium sold separately This can be used with 1 kit of 192152 (Keratinocyte Cell Growth Medium SingleQuots Supplements and Growth Factors) The full KGM Gold Calcium Free BulletKit consisting of basal medium and supplements can be purchased together using the catalog number 00195769 KGM Gold BulleKit is a robust and versatile serum-free medium formulation for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures