Learn More
Lonza Walkersville KBM Keratinocyte Basal Medium
Catalog No. 5000496437
Encompass
KBM Keratinocyte Basal Medium Calcium Free is a keratinocyte basal medium sold separately It can be used with 1 kit of CC-4131 (keratinocyte cell growth medium SingleQuotsTM Supplements and Growth Factors) The full KGMTM Calcium Free BulletKitTM consisting of basal medium and supplements can be purchased together using the catalog number CC-3112 KGMTM Keratinocyte Growth Medium Calcium Free has been optimized for the proliferation of keratinocytes in a serum-free and calcium-free environment Keratinocytes cannot survive in a completely calcium free system
Catalog No. 50-004-96437 Supplier Lonza Walkersville Supplier No. CC3104
KBM Keratinocyte Basal Medium Calcium Free is a keratinocyte basal medium sold separately It can be used with 1 kit of CC-4131 (keratinocyte cell growth medium SingleQuotsTM Supplements and Growth Factors) The full KGMTM Calcium Free BulletKitTM consisting of basal medium and supplements can be purchased together using the catalog number CC-3112 KGMTM Keratinocyte Growth Medium Calcium Free has been optimized for the proliferation of keratinocytes in a serum-free and calcium-free environment Keratinocytes cannot survive in a completely calcium free system