Learn More
Cayman Chemical KBP066trIfluoroACTAte sal 10mg
SDP
Catalog No. 502944890
Encompass_Preferred
A dual amylin and calcitonin receptor agonist; reduces body weight and food intake in rats fed a high-fat diet 50 UG/kg; prevents increases in fasting blood glucose levels in fa/fa Zucker diabetic fatty rats
Catalog No. 50-294-4890 Supplier Cayman Chemical Supplier No. 4133110 MG
A dual amylin and calcitonin receptor agonist; reduces body weight and food intake in rats fed a high-fat diet 50 UG/kg; prevents increases in fasting blood glucose levels in fa/fa Zucker diabetic fatty rats