Learn More
FIREBIRD OPTICS Potassium Bromide (KBr) Circular Optical Window- 25x4mm
SDP
Catalog No. NC1953401
Encompass_Preferred
Potassium Bromide (KBr) Circular Optical Window- 25x4mm• Wavelength range: 250nm-26µm •Surface quality: 60/40 • Clear aperture: 90% •Diameter tolerance: ±0.1mm •Thickness tolerance: ±0.2mm
Catalog No. NC1953401 Supplier FIREBIRD OPTICS Supplier No. KBR25X4
Potassium Bromide (KBr) Circular Optical Window- 25x4mm• Wavelength range: 250nm-26µm •Surface quality: 60/40 • Clear aperture: 90% •Diameter tolerance: ±0.1mm •Thickness tolerance: ±0.2mm