Learn More
Perkin Elmer US LLC KBr Window Kit for the Spectrum Two
Catalog No. 502111971
Encompass_Preferred
KBr has a slightly wider spectral range than NaCl but is more hygroscopic. Although able to provide good resistance to mechanical shock, KBr is easily cleaved.
Catalog No. 50-211-1971 Supplier Perkin Elmer US LLC Supplier No. L1600241
KBr has a slightly wider spectral range than NaCl but is more hygroscopic. Although able to provide good resistance to mechanical shock, KBr is easily cleaved.