Learn More
Aviva Systems Biology Corp KBTBD5 Antibody - middle region (ARP34663_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34663_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KBTBD5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KBTBD5. Alternate names: FLJ32015, MGC125350, SRYP, KBTBD5ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-55508