Learn More
Aviva Systems Biology Corp KBTBD8 Antibody - N-terminal region (ARP60271_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60271_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KBTBD8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human KBTBD8. Alternate names: FLJ57592, KIAA1842, TA-KRPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-991