Learn More
AdipoGen KBU2046
SDP
Catalog No. 502562355
Encompass_Preferred
Chemical. CAS 1143863-69-7. Formula C15H11FO2. MW 242.2. Highly selective metastasis inhibitor in vitro. Binds to and stabilizes Cdc37-HSP90beta heterocomplexes, consequently blocking its function and inhibiting cancer cell motility. Does not bind to Cdc37 or HSP90beta alone. Inhibits cancer metastasis in vivo in murine models of human prostate and breast cancer. Shown to exhibit a complete lack of cellular cytotoxicity and systemic toxicity.
Catalog No. 50-256-2355 Supplier AdipoGen Supplier No. AGCR10159M025
Chemical. CAS 1143863-69-7. Formula C15H11FO2. MW 242.2. Highly selective metastasis inhibitor in vitro. Binds to and stabilizes Cdc37-HSP90beta heterocomplexes, consequently blocking its function and inhibiting cancer cell motility. Does not bind to Cdc37 or HSP90beta alone. Inhibits cancer metastasis in vivo in murine models of human prostate and breast cancer. Shown to exhibit a complete lack of cellular cytotoxicity and systemic toxicity.