Learn More
Medchemexpress LLC HY-18777 10mg , KC7F2 CAS:927822-86-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1877710MG
Encompass_Preferred
HY-18777 10mg KC7F2 CAS: 927822-86-4 KC7F2 is a potent hypoxia inducible factor-1 (HIF-1) pathway inhibitor with an IC50 of 20 μM in LN229-HRE-AP cells, and with potential as a cancer therapy agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9384