Learn More
Selleck Chemical LLC KC7F2 20mg 927822-86-4
SDP
Catalog No. 501365459
Encompass_Preferred
KC7F2 is a selective HIF-1? translation inhibitor with IC50 of 20 ?M in a cell-based assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5459 Supplier Selleck Chemical LLC Supplier No. S794620MG
KC7F2 is a selective HIF-1? translation inhibitor with IC50 of 20 ?M in a cell-based assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.