Learn More
Medchemexpress LLC HY-131031 5mg , KCC-07 CAS:315702-75-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1310315MG
Encompass_Preferred
HY-131031 5mg KCC-07 CAS: 315702-75-1 KCC-07 is a potent, selective and brain-penetrant MBD2 (methyl-CpG-binding domain protein 2) inhibitor. KCC-07 prevents binding of MBD2 to methylated DNA and activates brain specific angiogenesis inhibitor 1 (BAI1) inducing anti-proliferative BAI1/p53/p21 signaling. Anticancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0705