Learn More
Medchemexpress LLC KCC009 | 744198-19-4 | 98.1% | 476.32 g/mol | C21H22BrN3O5 | 10MG
SDP
Catalog No. 5000238162
Encompass_Preferred
KCC009 is a small-molecule transglutaminase 2 (TG2) inhibitor used for laboratory research. It has been shown to induce p53-independent radiosensitization and to disrupt fibronectin assembly in the extracellular matrix, supporting cellular and in vivo studies of TG2-related biology. Follow handling, solvent, and storage instructions in the SDS for experimental use.
Catalog No. 50-002-38162 Supplier Medchemexpress LLC Supplier No. HY12329010MG
KCC009 is a small-molecule transglutaminase 2 (TG2) inhibitor used for laboratory research. It has been shown to induce p53-independent radiosensitization and to disrupt fibronectin assembly in the extracellular matrix, supporting cellular and in vivo studies of TG2-related biology. Follow handling, solvent, and storage instructions in the SDS for experimental use.