Learn More
Medchemexpress LLC KCC009 | 744198-19-4 | MFCD12032123 | 98.1% | 476.32 g/mol | C21H22BrN3O5 | 5MG
SDP
Catalog No. 5000238307
Encompass_Preferred
KCC009 is a small-molecule transglutaminase 2 (TG2) inhibitor supplied for research use. It has been reported to induce p53-independent radiosensitization and to disrupt fibronectin assembly in the extracellular matrix. The compound is identified by CAS 744198-19-4, has a molecular weight of 476.32 g/mol, chemical formula C21H22BrN3O5, and is provided as a solid with reported purity of 98.1%.
  • Inhibits transglutaminase 2 activity.
  • Induces p53-independent radiosensitization in cancer cells.
  • Disrupts fibronectin assembly in the extracellular matrix.
  • Provided as a solid suitable for in vitro research.
  • Includes a product data sheet and safety data sheet for handling information.
Catalog No. 50-002-38307 Supplier Medchemexpress LLC Supplier No. HY1232905MG
KCC009 is a small-molecule transglutaminase 2 (TG2) inhibitor supplied for research use. It has been reported to induce p53-independent radiosensitization and to disrupt fibronectin assembly in the extracellular matrix. The compound is identified by CAS 744198-19-4, has a molecular weight of 476.32 g/mol, chemical formula C21H22BrN3O5, and is provided as a solid with reported purity of 98.1%.
  • Inhibits transglutaminase 2 activity.
  • Induces p53-independent radiosensitization in cancer cells.
  • Disrupts fibronectin assembly in the extracellular matrix.
  • Provided as a solid suitable for in vitro research.
  • Includes a product data sheet and safety data sheet for handling information.