Learn More
Medchemexpress LLC KCC2 blocker 1 | 1228439-36-8 | 98.2% | 415.50 | 1 ML
SDP
Catalog No. 5000386904
Encompass_Preferred
KCC2 blocker 1 is an orally active and selective K+-Cl- cotransporter KCC2 blocker with an IC50 of 1 μM. It is identified as a benzyl prolinate. This compound selectively inhibits KCC2, making it a valuable tool for neuronal signaling research.
  • Orally active.
  • Selective K+-Cl- cotransporter KCC2 blocker.
  • Exhibits an IC50 of 1 μM for KCC2.
  • Classified as a benzyl prolinate.
  • Demonstrates antagonist activity against NKCC1 in MDCK cells.
  • Applicable for research purposes.
Catalog No. 50-003-86904 Supplier Medchemexpress LLC Supplier No. HY1817210MM/1ML
KCC2 blocker 1 is an orally active and selective K+-Cl- cotransporter KCC2 blocker with an IC50 of 1 μM. It is identified as a benzyl prolinate. This compound selectively inhibits KCC2, making it a valuable tool for neuronal signaling research.
  • Orally active.
  • Selective K+-Cl- cotransporter KCC2 blocker.
  • Exhibits an IC50 of 1 μM for KCC2.
  • Classified as a benzyl prolinate.
  • Demonstrates antagonist activity against NKCC1 in MDCK cells.
  • Applicable for research purposes.