Learn More
Medchemexpress LLC KCC2 blocker 1 | 1228439-36-8 | 98.2% | 415.50 | 5 MG
SDP
Catalog No. 5000387253
Encompass_Preferred
KCC2 blocker 1 is an orally active and selective K+-Cl- cotransporter KCC2 blocker with an IC50 of 1 μM. It is a benzyl prolinate.
  • Orally active and selective K+-Cl- cotransporter KCC2 blocker
  • IC50 of 1 μM (KCC2)
  • Benzyl prolinate
  • It has a t1/2 of 0.3 hours, a CL of 26 mL/min/kg, a C max of 457 ng/mL and a AUC of 726 ng·h/mL for male Wistar rats.
Catalog No. 50-003-87253 Supplier Medchemexpress LLC Supplier No. HY181725MG
KCC2 blocker 1 is an orally active and selective K+-Cl- cotransporter KCC2 blocker with an IC50 of 1 μM. It is a benzyl prolinate.
  • Orally active and selective K+-Cl- cotransporter KCC2 blocker
  • IC50 of 1 μM (KCC2)
  • Benzyl prolinate
  • It has a t1/2 of 0.3 hours, a CL of 26 mL/min/kg, a C max of 457 ng/mL and a AUC of 726 ng·h/mL for male Wistar rats.