Learn More
Medchemexpress LLC (+)-KCC2 blocker 1 | 1228439-71-1 | 98.7% | 415.50 | 1 MG
SDP
Catalog No. 5000387209
Encompass_Preferred
(+)-KCC2 blocker 1 is a selective K+-Cl- cotransporter KCC2 blocker with an IC50 of 0.4 μM. It is a benzyl prolinate and an enantiomer of KCC2 blocker 1. It is intended for research use only and is not sold to patients.
  • Selective K+-Cl- cotransporter KCC2 blocker
  • IC50 of 0.4 μM for KCC2
  • Benzyl prolinate
  • Enantiomer of KCC2 blocker 1
Catalog No. 50-003-87209 Supplier Medchemexpress LLC Supplier No. HY18172A1MG
(+)-KCC2 blocker 1 is a selective K+-Cl- cotransporter KCC2 blocker with an IC50 of 0.4 μM. It is a benzyl prolinate and an enantiomer of KCC2 blocker 1. It is intended for research use only and is not sold to patients.
  • Selective K+-Cl- cotransporter KCC2 blocker
  • IC50 of 0.4 μM for KCC2
  • Benzyl prolinate
  • Enantiomer of KCC2 blocker 1