Learn More
TARGETMOL CHEMICALS INC KCC2 BLOCKER 1 5MG
SDP
Catalog No. 502581447
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (+)-KCC2 blocker 1 is a selective blocker of KCC2 (IC50 = 0.4 uM). purity: 98%
Catalog No. 50-258-1447 Supplier TARGETMOL CHEMICALS INC Supplier No. T125045MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (+)-KCC2 blocker 1 is a selective blocker of KCC2 (IC50 = 0.4 uM). purity: 98%