Learn More
Cayman Chemical KCC2PhosphoSR940 Polyclo 100uL
SDP
Catalog No. 502938496
Encompass_Preferred
An esterified form of nonanoic acid found as a volatile component following thermal oxidation of conjugated linoleic acid methyl ester cytotoxic to A549 lung carcinoma cells (LC50 104 09 Ug/ml) enhances the penetration of minoxidil into isolated hamster ventral ear skin when applied at a 10 concentration ex vivo a substrate for E coli AlkBGT
Catalog No. 50-293-8496 Supplier Cayman Chemical Supplier No. 29291100 UL
An esterified form of nonanoic acid found as a volatile component following thermal oxidation of conjugated linoleic acid methyl ester cytotoxic to A549 lung carcinoma cells (LC50 104 09 Ug/ml) enhances the penetration of minoxidil into isolated hamster ventral ear skin when applied at a 10 concentration ex vivo a substrate for E coli AlkBGT