Learn More
TARGETMOL CHEMICALS INC KCL-286 5MG
SDP
Catalog No. 502879760
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. KCL-286 is an orally available and potent retinoic acid receptor beta agonist for the amelioration of spinal cord injury (SCI). Purity 99.93%
Catalog No. 50-287-9760 Supplier TARGETMOL CHEMICALS INC Supplier No. T785755MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. KCL-286 is an orally available and potent retinoic acid receptor beta agonist for the amelioration of spinal cord injury (SCI). Purity 99.93%