Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Electrolyte solution (KCl 3M), 50mL

Catalog No. 501998015 Shop All Hach Company Products
Encompass

Electrolyte Solution (3 M KCl), 50 mL Bottle

Catalog No. 50-199-8015 Supplier Hach Company Supplier No. LZW9509.99
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Electrolyte Solution (3 M KCl), 50 mL Bottle

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.