Learn More
Broadley James Corporation pH Electrode Storage Solution, 500 mL
Catalog No. NC9055961
Encompass
Broadley-James now offers electrode storage solution. Potassium Chloride (KCl) solution is the first choice for storing FermProbes. If this is not available, buffer solution can be used. NEVER store pH electrodes in deionized (DI) water.500 mL
Catalog No. NC9055961 Supplier Broadley James Corporation Supplier No. AS3120C200500
Broadley-James now offers electrode storage solution. Potassium Chloride (KCl) solution is the first choice for storing FermProbes. If this is not available, buffer solution can be used. NEVER store pH electrodes in deionized (DI) water.500 mL