Learn More
International Crystal SpectroGrade™ KCl Powder, 100g
Catalog No. NC0227451
Encompass_Preferred
Spectrograde™ KCl powder makes IR pellets of extremely high quality with low moisture content and low impurity content. Spectrograde™ KCl comes directly from ICL’s own KCl ingot stock. This powder is ideal for producing KCl pellets and a 100gm bottle will yield approximately 250 7mm pellets.
Catalog No. NC0227451 Supplier International Crystal Supplier No. 00114129
Spectrograde™ KCl powder makes IR pellets of extremely high quality with low moisture content and low impurity content. Spectrograde™ KCl comes directly from ICL’s own KCl ingot stock. This powder is ideal for producing KCl pellets and a 100gm bottle will yield approximately 250 7mm pellets.