Learn More
Aviva Systems Biology Corp KCNA10 Antibody - middle region (ARP35178_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35178_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNA10. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human KCNA10. Alternate names: Kcn1, Kv1.8ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-55662