Learn More
Aviva Systems Biology Corp KCNA10 Antibody - N-terminal region (ARP35177_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35177_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNA10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCNA10. Alternate names: Kcn1, Kv1.8ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55661