Learn More
Aviva Systems Biology Corp KCNAB2 Antibody - middle region (ARP37678_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37678_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNAB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNAB2. Alternate names: AKR6A5, KCNA2B, HKvbeta2, KV-BETA-2, HKvbeta2.1, HKvbeta2.2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-56392