Learn More
Aviva Systems Biology Corp KCNC3 Antibody - middle region (ARP35158_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35158_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNC3. Alternate names: KSHIIID, KV3.3, SCA13ApplicationWB, IHC. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55655