Learn More
Aviva Systems Biology Corp KCNC4 Antibody - middle region (ARP35161_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35161_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNC4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNC4. Alternate names: HKSHIIIC, KSHIIIC, KV3.4, MGC126818, C1orf30ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55656