Learn More
Aviva Systems Biology Corp Kcnd1 Antibody - N-terminal region (ARP35163_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35163_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Kcnd1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1110037K09Rik, Kca2-1, Kv4.1, Shal, mShal1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-448