Learn More
Aviva Systems Biology Corp KCND3 Antibody - middle region (ARP35467_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35467_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCND3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCND3. Alternate names: KCND3L, KCND3S, KSHIVB, KV4.3, MGC142035, MGC142037ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55747