Learn More
Aviva Systems Biology Corp KCNG1 Antibody - N-terminal region (ARP35470_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35470_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCNG1. Alternate names: K13, KCNG, KV6.1, MGC12878, kH2ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-55749