Learn More
Aviva Systems Biology Corp KCNG4 Antibody - N-terminal region (ARP35473_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35473_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNG4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCNG4. Alternate names: KV6.3, KV6.4, MGC129609, MGC4558ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55750