Learn More
Aviva Systems Biology Corp KCNH2 Antibody - C-terminal region (ARP35450_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35450_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNH2. Alternate names: ERG1, HERG, HERG1, Kv11.1, LQT2, SQT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55737