Learn More
Aviva Systems Biology Corp KCNH3 Antibody - middle region (ARP37688_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37688_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNH3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNH3. Alternate names: BEC1, ELK2, KIAA1282, Kv12.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56395