Learn More
Aviva Systems Biology Corp KCNH3 Antibody - N-terminal region (ARP35213_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35213_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNH3. It was validated on Western Blot. . Alternate names: BEC1, ELK2, KIAA1282, Kv12.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-454