Learn More
Aviva Systems Biology Corp KCNH6 Antibody - N-terminal region (ARP35363_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35363_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNH6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human KCNH6. Alternate names: ERG2, HERG2, Kv11.2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55712