Learn More
Aviva Systems Biology Corp KCNH6 Antibody - middle region (ARP35481_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35481_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNH6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNH6. Alternate names: ERG2, HERG2, Kv11.2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55753