Learn More
Aviva Systems Biology Corp Kcnh8 Antibody - C-terminal region (ARP35410_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35410_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Kcnh8. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: C130090D05Rik, ELK, ELK1, ELK3, Kv12.1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-462