Learn More
Aviva Systems Biology Corp KCNIP1 Antibody - N-terminal region (ARP35246_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35246_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNIP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human KCNIP1. Alternate names: VABP, KCHIP1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55680